Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097098.2 | XP_014952584.2 | 2 - 3 | 743 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |