Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002824.4 | XP_004002873.2 | 5 - 6 | 655 | 77 | |
XM_004002825.4 | XP_004002874.3 | 6 - 7 | 1280 | 77 |