Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096190.2 | XP_014951676.2 | 4 - 5 | 2893 | 122 | |
XM_027970507.1 | XP_027826308.1 | 4 - 5 | 451 | 122 | ||
XM_027970508.1 | XP_027826309.1 | 4 - 5 | 451 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |