Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015228.4 | XP_004015277.1 | 2 - 3 | 875 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |