Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978560.1 | XP_027834361.1 | 3 - 4 | 2337 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |