Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100319.2 | XP_014955805.2 | 7 - 8 | 1518 | 79 | |
XM_027978798.1 | XP_027834599.1 | 6 - 7 | 1155 | 79 | ||
XM_027978799.1 | XP_027834600.1 | 7 - 8 | 1046 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |