Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970115.1 | XP_027825916.1 | 6 - 7 | 1579 | 63 | |
XM_027970116.1 | XP_027825917.1 | 6 - 7 | 1569 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |