Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003214.4 | XP_004003263.2 | 1 - 2 | 404 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |