Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958945.1 | XP_027814746.1 | 4 - 5 | 603 | 57 | |
XM_027958946.1 | XP_027814747.1 | 4 - 5 | 600 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |