Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009526.4 | XP_004009575.3 | 3 - 4 | 291 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |