Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097958.2 | XP_014953444.2 | 2 - 3 | 308 | 79 | |
XM_027973760.1 | XP_027829561.1 | 2 - 3 | 1177 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |