Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966021.1 | XP_027821822.1 | 5 - 6 | 689 | 68 | |
XM_027966022.1 | XP_027821823.1 | 5 - 6 | 775 | 68 |