Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093423.2 | XP_014948909.1 | 4 - 5 | 314 | 100 | |
XM_015093424.2 | XP_014948910.1 | 3 - 4 | 237 | 100 |