Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186058.3 | XP_012041448.1 | 5 - 6 | 1804 | 67 | |
XM_015098764.2 | XP_014954250.1 | 4 - 5 | 1756 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |