Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002786.3 | XP_004002835.1 | 4 - 5 | 667 | 62 | |
XM_012186695.2 | XP_012042085.1 | 5 - 6 | 418 | 62 | ||
XM_012186700.2 | XP_012042090.1 | 4 - 5 | 396 | 62 | ||
XM_012186706.3 | XP_012042096.1 | 4 - 5 | 309 | 62 | ||
XM_012186712.2 | XP_012042102.1 | 5 - 6 | 723 | 62 | ||
XM_027978512.1 | XP_027834313.1 | 5 - 6 | 450 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |