Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012987.4 | XP_004013036.3 | 5 - 6 | 525 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |