Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013114.4 | XP_004013163.2 | 8 - 9 | 2437 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |