Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095281.2 | XP_011950671.2 | 17 - 18 | 2134 | 72 | |
XM_012095282.3 | XP_011950672.2 | 17 - 18 | 2098 | 72 | ||
XM_012095283.2 | XP_011950673.2 | 15 - 16 | 1760 | 72 | ||
XM_012095284.2 | XP_011950674.2 | 14 - 15 | 1688 | 72 |