Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978187.1 | XP_027833988.1 | 3 - 4 | 395 | 80 | |
XM_027978188.1 | XP_027833989.1 | 3 - 4 | 359 | 80 | ||
XM_027978189.1 | XP_027833990.1 | 3 - 4 | 298 | 80 | ||
XM_027978190.1 | XP_027833991.1 | 3 - 4 | 262 | 80 | ||
XM_027978191.1 | XP_027833992.1 | 3 - 4 | 401 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |