Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977938.1 | XP_027833739.1 | 3 - 4 | 747 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Ovis aries |