Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965962.1 | XP_027821763.1 | 2 - 3 | 279 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |