Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012585.4 | XP_004012634.2 | 5 - 6 | 813 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |