Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958084.1 | XP_027813885.1 | 2 - 3 | 1188 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |