Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103821.3 | XP_011959211.2 | 3 - 4 | 1112 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |