Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012174980.3 | XP_012030370.2 | 2 - 3 | 518 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |