Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010798.4 | XP_004010847.1 | 3 - 4 | 473 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |