Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093019.2 | XP_014948505.1 | 3 - 4 | 1285 | 74 | |
XM_027964174.1 | XP_027819975.1 | 3 - 4 | 363 | 74 | ||
XM_027964175.1 | XP_027819976.1 | 4 - 5 | 1700 | 74 | ||
XM_027964176.1 | XP_027819977.1 | 2 - 3 | 116 | 74 | ||
XM_027964178.1 | XP_027819979.1 | 2 - 3 | 218 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |