Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012772.4 | XP_004012821.1 | 5 - 6 | 863 | 75 | |
XM_015098714.2 | XP_014954200.1 | 5 - 6 | 788 | 75 | ||
XM_027974709.1 | XP_027830510.1 | 5 - 6 | 548 | 75 |