Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095867.2 | XP_014951353.1 | 6 - 7 | 1151 | 60 | |
XM_015095868.2 | XP_014951354.1 | 5 - 6 | 1025 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |