Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958398.1 | XP_027814199.1 | 3 - 4 | 636 | 78 | |
XM_027958399.1 | XP_027814200.1 | 3 - 4 | 515 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |