Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969218.1 | XP_027825019.1 | 3 - 4 | 360 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |