Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093613.2 | XP_014949099.2 | 3 - 4 | 397 | 75 | |
XM_015093614.2 | XP_014949100.2 | 3 - 4 | 397 | 75 | ||
XM_015093615.2 | XP_014949101.2 | 3 - 4 | 397 | 75 |