Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187544.3 | XP_012042934.1 | 5 - 6 | 846 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |