Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014323.4 | XP_004014372.1 | 8 - 9 | 999 | 119 | |
XM_012188608.3 | XP_012043998.1 | 7 - 8 | 831 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |