Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958971.1 | XP_027814772.1 | 7 - 8 | 1474 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |