Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962400.1 | XP_027818201.1 | 1 - 2 | 1433 | 69 | |
XM_027962401.1 | XP_027818202.1 | 2 - 3 | 1275 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |