Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107841.3 | XP_011963231.3 | 1 | 315 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |