Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971433.1 | XP_027827234.1 | 6 - 7 | 1230 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |