Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978682.1 | XP_027834483.1 | 7 - 8 | 917 | 76 | |
XM_027978692.1 | XP_027834493.1 | 7 - 8 | 982 | 76 | ||
GenBank mRNA | AF057717.1 | - | 3 - 4 | 452 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |