Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969311.1 | XP_027825112.1 | 8 - 9 | 1546 | 102 | |
XM_027969322.1 | XP_027825123.1 | 8 - 9 | 1538 | 102 | ||
XM_027969328.1 | XP_027825129.1 | 8 - 9 | 1551 | 102 | ||
XM_027969331.1 | XP_027825132.1 | 8 - 9 | 1520 | 102 | ||
XM_027969339.1 | XP_027825140.1 | 8 - 9 | 1512 | 102 | ||
XM_027969345.1 | XP_027825146.1 | 8 - 9 | 1543 | 102 | ||
XM_027969351.1 | XP_027825152.1 | 7 - 8 | 1463 | 102 | ||
XM_027969355.1 | XP_027825156.1 | 7 - 8 | 1116 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |