Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014577.4 | XP_004014626.1 | 14 - 15 | 1599 | 69 | |
XM_027977208.1 | XP_027833009.1 | 13 - 14 | 1289 | 69 | ||
XM_027977209.1 | XP_027833010.1 | 13 - 14 | 1286 | 69 | ||
XM_027977211.1 | XP_027833012.1 | 12 - 13 | 1199 | 69 | ||
XM_027977214.1 | XP_027833015.1 | 13 - 14 | 1452 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |