Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103585.3 | XP_011958975.2 | 3 - 4 | 926 | 87 | |
XM_012103586.3 | XP_011958976.2 | 3 - 4 | 1258 | 87 |