Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958150.1 | XP_027813951.1 | 4 - 5 | 2209 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |