Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114111203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962888.1 | XP_027818689.1 | 1 | 318 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |