Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009590.4 | XP_004009639.2 | 1 - 2 | 220 | 58 | |
XM_012179415.3 | XP_012034805.2 | 1 - 2 | 220 | 58 | ||
XM_012179416.3 | XP_012034806.2 | 1 - 2 | 218 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |