Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP1BA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185590.3 | XP_012040980.2 | 1 - 2 | 683 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |