Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979950.1 | XP_027835751.1 | 1 - 2 | 48 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |