Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964000.1 | XP_027819801.1 | 5 - 6 | 1590 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Ovis aries |