Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011387.3 | XP_004011436.1 | 2 - 3 | 253 | 117 | |
XM_027972506.1 | XP_027828307.1 | 2 - 3 | 253 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |