Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962639.1 | XP_027818440.1 | 3 - 4 | 354 | 90 | |
XM_027962640.1 | XP_027818441.1 | 3 - 4 | 354 | 90 | ||
XM_027962641.1 | XP_027818442.1 | 3 - 4 | 354 | 90 | ||
XM_027962642.1 | XP_027818443.1 | 2 - 3 | 260 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |